missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD28 Protein
A cDNA sequence encoding the CD28 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11322-10ug
Additional Details : Weight : 0.01000kg
Specifications
940 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPNKILVKQ SPMLVAYDNA VNLSCKYSYN LFSREFRASL HKGLDSAVEV CVVYGNYSQQ LQVYSKTGFN CDGKLGNESV TFYLQNLYVN QTDIYFCKIE VMYPPPYLDN EKSNGTIIHV KGKHLCPSPL FPGPSKPLEP KSCDKTHTCP PCPAPELLGG PSVFLFPPKP KDTLMISRTP EVTCVVVDVS HEDPEVKFNW YVDGVEVHNA KTKPREEQYN STYRVVSVLT VLHQDWLNGK EYKCKVSNKA LPAPIEKTIS KAKGQPREPQ VYTLPPSRDE LTKNQVSLTC LVKGFYPSDI AVEWESNGQP ENNYKTTPPV LDSDGSFFLY SKLTVDKSRW QQGNVFSCSV MHEALHNHYT QKSLSLSPGK HHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
CD28 Protein | |
Research Use Only | |
CD28 | |
Human | |
His | |
CD28 protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |