missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300A Protein
A cDNA sequence encoding the CD300A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11324-2ug
Additional Details : Weight : 0.01000kg
Specifications
11314 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CD300A | |
Human | |
His | |
The CD300A solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |
CD300A Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLSKCRTV AGPVGGSLSV QCPYEKEHRT LNKYWCRPPQ IFLCDKIVET KGSAGKRNGR VSIRDSPANL SFTVTLENLT EEDAGTYWCG VDTPWLRDFH DPVVEVEVSV FPAS |