missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD300LF / LMIR3 Protein

A DNA sequence encoding the Homo sapiens (Human) CD300LF / LMIR3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7829-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051041

  • 2285.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001276011.1
CD300LF / LMIR3 Protein
Research Use Only
CD300LF
Recombinant Protein
E. coli
TQITGPTTVN GLERGSLTVQ CVYRSGWETY LKWWCRGAIW RDCKILVKTS GSEQEVKRDR VSIKDNQKNR TFTVTMEDLM KTDADTYWCG IEKTGNDLGV TVQVTIDPAP VTQEETSSSP TLTGHHLDNR HKLLKLS
Greater than 90% as determined by SDS-PAGE.
146722
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers