missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD34 Protein

A cDNA sequence encoding the CD34 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11328-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936878

  • 397.00 € / 5µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

947
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
MLVRRGARAG PRMPRGWTAL CLLSLLPSGF MSLDNNGTAT PELPTQGTFS NVSTNVSYQE TTTPSTLGST SLHPVSQHGN EATTNITETT VKFTSTSVIT SVYGNTNSSV QSQTSVISTV FTTPANVSTP ETTLKPSLSP GNVSDLSTTS TSLATSPTKP YTSSSPILSD IKAEIKCSGI REVKLTQGIC LEQNKTSSCA EFKKDRGEGL ARVLCGEEQA DADAGAQVCS LLLAQSEVRP QCLLLVLANR TEISSKLQLM KKHQSDLKKL GILDFTEQDV ASHQSYSQKT LIALVTSGAL LAVLGITGYF LMNRRSWSPT GERLGEDPYY TENGGGQGYS SGPGTSPEAQ GKASVNRGAQ ENGTGQATSR NGHSARQHVV ADTEL
CD34 Protein
Research Use Only
CD34
Human
Untagged
CD34 in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers