missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD38 Protein
A cDNA sequence encoding the CD38 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 952 |
---|---|
Name | CD38 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD38 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986888
|
enQuireBio™
QP11331-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966888
|
enQuireBio™
QP11331-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976888
|
enQuireBio™
QP11331-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
952 | |
Research Use Only | |
CD38 | |
Human | |
His | |
CD38 protein solution (1 mg/ml) contains 100mM NaCl and 10% glycerol. |
CD38 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPEFVPRWR QQWSGPGTTK RFPETVLARC VKYTEIHPEM RHVDCQSVWD AFKGAFISKH PCNITEEDYQ PLMKLGTQTV PCNKILLWSR IKDLAHQFTQ VQRDMFTLED TLLGYLADDL TWCGEFNTSK INYQSCPDWR KDCSNNPVSV FWKTVSRRFA EAACDVVHVM LNGSRSKIFD KNSTFGSVEV HNLQPEKVQT LEAWVIHGGR EDSRDLCQDP TIKELESIIS KRNIQFSCKN IYRPDKFLQC VKNPEDSSCT SEIHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |