missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD5 Protein

A cDNA sequence encoding the CD5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11338-HIS-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956918

  • 131.00 € / 2µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

921
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSRLSWYDP DFQARLTRSN SKCQGQLEVY LKDGWHMVCS QSWGRSSKQW EDPSQASKVC QRLNCGVPLS LGPFLVTYTP QSSIICYGQL GSFSNCSHSR NDMCHSLGLT CLEPQKTTPP TTRPPPTTTP EPTAPPRLQL VAQSGGQHCA GVVEFYSGSL GGTISYEAQD KTQDLENFLC NNLQCGSFLK HLPETEAGRA QDPGEPREHQ PLPIQWKIQN SSCTSLEHCF RKIKPQKSGR VLALLCSGFQ PKVQSRLVGG SSICEGTVEV RQGAQWAALC DSSSARSSLR WEEVCREQQC GSVNSYRVLD AGDPTSRGLF CPHQKLSQCH ELWERNSYCK KVFVTCQDPN P
Greater than 80% as determined by SDS-PAGE.
CD5 Protein
Research Use Only
CD5
Human
His
The CD5 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers