missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD59 / CD59A / MAC-IP Protein

A DNA sequence encoding the Homo sapiens (Human) CD59 / CD59A / MAC-IP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5791-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937789

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000602.1
CD59 / CD59A / MAC-IP Protein
Research Use Only
CD59
Recombinant Protein
Yeast
LQCYNCPNPT ADCKTAVNCS SDFDACLITK AGLQVYNKCW KFEHCNFNDV TTRLRENELT YYCCKKDLCN FNEQLEN
Greater than 90% as determined by SDS-PAGE.
966
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers