missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD59 / CD59A / MAC-IP Protein
A DNA sequence encoding the Homo sapiens (Human) CD59 / CD59A / MAC-IP, was expressed in the hosts and tags indicated.
252.00 € - 2645.00 €
Specifications
Accession Number | NP_000602.1 |
---|---|
Gene ID (Entrez) | 966 |
Name | CD59 / CD59A / MAC-IP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15917789
|
enQuireBio™
QP5791-YE-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907789
|
enQuireBio™
QP5791-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927789
|
enQuireBio™
QP5791-YE-1MG |
1 mg |
2645.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937789
|
enQuireBio™
QP5791-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15947789
|
enQuireBio™
QP5791-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957789
|
enQuireBio™
QP5791-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000602.1 | |
CD59 / CD59A / MAC-IP Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
966 | |
Research Use Only | |
CD59 | |
Recombinant Protein | |
Yeast | |
LQCYNCPNPT ADCKTAVNCS SDFDACLITK AGLQVYNKCW KFEHCNFNDV TTRLRENELT YYCCKKDLCN FNEQLEN | |
Greater than 90% as determined by SDS-PAGE. |