missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD80 Protein

A cDNA sequence encoding the CD80 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11350-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986968

  • 131.00 € / 1µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD80 Protein
Research Use Only
Recombinant Protein
Baculovirus
VIHVTKEVKE VATLSCGHNV SVEELAQTRI YWQKEKKMVL TMMSGDMNIW PEYKNRTIFD ITNNLSIVIL ALRPSDEGTY ECVVLKYEKD AFKREHLAEV TLSVKADFPT PSISDFEIPT SNIRRIICST SGGFPEPHLS WLENGEELNA INTTVSQDPE TELYAVSSKL DFNMTTNHSF MCLIKYGHLR VNQTFNWNTT KQEHFPDNLE HHHHHH
Greater than 95.0% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CD80 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers