missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD80 Protein
A cDNA sequence encoding the CD80 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11350-50ug
Additional Details : Weight : 0.01000kg
Specifications
CD80 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VIHVTKEVKE VATLSCGHNV SVEELAQTRI YWQKEKKMVL TMMSGDMNIW PEYKNRTIFD ITNNLSIVIL ALRPSDEGTY ECVVLKYEKD AFKREHLAEV TLSVKADFPT PSISDFEIPT SNIRRIICST SGGFPEPHLS WLENGEELNA INTTVSQDPE TELYAVSSKL DFNMTTNHSF MCLIKYGHLR VNQTFNWNTT KQEHFPDNLE HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CD80 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |