missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD83 Protein
A cDNA sequence encoding the CD83 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11351-1mg
Specifications
9308 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTPEVKVA CSEDVDLPCT APWDPQVPYT VSWVKLLEGG EERMETPQED HLRGQHYHQK GQNGSFDAPN ERPYSLKIRN TTSCNSGTYR CTLQDPDGQR NLSGKVILRV TGCPAQRKEE TFKKYRAE | |
Greater than 90% as determined by SDS-PAGE. |
CD83 Protein | |
Research Use Only | |
CD83 | |
Human | |
His | |
CD83 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 30% glycerol. |