missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD96 Protein
A cDNA sequence encoding the CD96 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 10225 |
---|---|
Name | CD96 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD96 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907008
|
enQuireBio™
QP11358-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986998
|
enQuireBio™
QP11358-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996998
|
enQuireBio™
QP11358-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10225 | |
Research Use Only | |
CD96 | |
Human | |
His | |
CD96 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
CD96 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
VWEKTVNTEE NVYATLGSDV NLTCQTQTVG FFVQMQWSKV TNKIDLIAVY HPQYGFYCAY GRPCESLVTF TETPENGSKW TLHLRNMSCS VSGRYECMLV LYPEGIQTKI YNLLIQTHVT ADEWNSNHTI EIEINQTLEI PCFQNSSSKI SSEFTYAWSV EDNGTQETLI SQNHLISNST LLKDRVKLGT DYRLHLSPVQ IFDDGRKFSC HIRVGPNKIL RSSTTVKVFA KPEIPVIVEN NSTDVLVERR FTCLLKNVFP KANITWFIDG SFLHDEKEGI YITNEERKGK DGFLELKSVL TRVHSNKPAQ SDNLTIWCMA LSPVPGNKVW NISSEKITFL LGSEISSTDP PLSVTESTLD TQPSPASSVS PARYPATSSV TLVDVSALRP NTTPQPSNSS MTTRGFNYPW TSSGTDTKKS VSRIPSETYS SSPSGAGSTL HDNVFTSTAR AFSEVPTTAN GSTKTNHVHI TGIVVNKPKD GMLEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |