missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDCA8 Protein
A cDNA sequence encoding the CDCA8 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | CDCA8 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957038
|
enQuireBio™
QP11366-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937038
|
enQuireBio™
QP11366-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947038
|
enQuireBio™
QP11366-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CDCA8 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CDCA8 solution (0.25 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 50% glycerol, 1mM DTT, 1mM EDTA and 0.2M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRKLASFLKD FDREVEIRIK QIESDRQNLL KEVDNLYNIE ILRLPKALRE MNWLDYFALG GNKQALEEAA TADLDITEIN KLTAEAIQTP LKSAKTRKVI QVDEMIVEEE EEEENERKNL QTARVKRCPP SKKRTQSIQG KGKGKRSSRA NTVTPAVGRLEVSMVKPTPG LTPRFDSRVF KTPGLRTPAA GERIYNISGN GSPLADSKEI FLTVPVGGGE SLRLLASDLQ RHSIAQLDPE ALGNIKKLSN RLAQICSSIR THK | |
Greater than 90.0% as determined by SDS-PAGE. |