missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CDK1 Protein

A cDNA sequence encoding the CDK1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11372-HIS-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15917058

  • 203.00 € / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

983
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MEDYTKIEKI GEGTYGVVYK GRHKTTGQVV AMKKIRLESE EEGVPSTAIR EISLLKELRH PNIVSLQDVL MQDSRLYLIF EFLSMDLKKY LDSIPPGQYM DSSLVKSYLY QILQGIVFCH SRRVLHRDLK PQNLLIDDKG TIKLADFGLA RAFGIPIRVY THEVVTLWYR SPEVLLGSAR YSTPVDIWSI GTIFAELATK KPLFHGDSEI DQLFRIFRAL GTPNNEVWPE VESLQDYKNT FPKWKPGSLA SHVKNLDENG LDLLSKMLIY DPAKRISGKM ALNHPYFNDL DNQIKKMLEH HHHHH
Greater than 95.0% as determined by SDS-PAGE.
CDK1 Protein
Research Use Only
CDK1
Human
His
CDK1 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4), 30% glycerol, 2mM DTT and 0.1mM PMSF.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers