missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK1 Protein
A cDNA sequence encoding the CDK1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11372-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
983 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MEDYTKIEKI GEGTYGVVYK GRHKTTGQVV AMKKIRLESE EEGVPSTAIR EISLLKELRH PNIVSLQDVL MQDSRLYLIF EFLSMDLKKY LDSIPPGQYM DSSLVKSYLY QILQGIVFCH SRRVLHRDLK PQNLLIDDKG TIKLADFGLA RAFGIPIRVY THEVVTLWYR SPEVLLGSAR YSTPVDIWSI GTIFAELATK KPLFHGDSEI DQLFRIFRAL GTPNNEVWPE VESLQDYKNT FPKWKPGSLA SHVKNLDENG LDLLSKMLIY DPAKRISGKM ALNHPYFNDL DNQIKKMLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CDK1 Protein | |
Research Use Only | |
CDK1 | |
Human | |
His | |
CDK1 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4), 30% glycerol, 2mM DTT and 0.1mM PMSF. |