missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK16 Protein
A cDNA sequence encoding the CDK16 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11373-20ug
Additional Details : Weight : 0.01000kg
Specifications
CDK16 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGFGKLET YIKLDKLGEG TYATVYKGKS KLTDNLVALK EIRLEHEEGA PCTAIREVSL LKDLKHANIV TLHDIIHTEK SLTLVFEYLD KDLKQYLDDC GNIINMHNVK LFLFQLLRGL AYCHRQKVLH RDLKPQNLLI NERGELKLAD FGLARAKSIP TKTYSNEVVT LWYRPPDILL GSTDYSTQID MWGVGCIFYE MATGRPLFPG STVEEQLHFI FRILGTPTEE TWPGILSNEE FKTYNYPKYR AEALLSHAPR LDSDGADLLT KLLQFEGRNR ISAEDAMKHP FFLSLGERIH KLPDTTSIFA LKEIQLQKEA SLRSSSMPDS GRPAFRVVDT EF | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CDK16 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |