missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human cDNA FLJ56323 Protein
A DNA sequence encoding the Homo sapiens (Human) cDNA FLJ56323, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8731-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002403.2 | |
cDNA FLJ56323 Protein | |
Research Use Only | |
MGMT | |
Recombinant Protein | |
E. coli | |
QPAPLERFAS RRPQVLAVRT VCDLVLGKMD KDCEMKRTTL DSPLGKLELS GCEQGLHEIK LLGKGTSAAD AVEVPAPAAV LGGPEPLMQC TAWLNAYFHQ PEAIEEFPVP ALHHPVFQQE SFTRQVLWKL LKVVKFGEVI SYQQLAALAG NPKAARAVGG AMRGNPVPIL IPCHRVVCSS GAVGNYSGGL AVKEWLLAHE GHRLGKPGLG GSSGLAGAWL KGAGATSGSP PAGRN | |
Greater than 85.0% as determined by SDS-PAGE. |
4255 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |