missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human cDNA FLJ56323 Protein
A DNA sequence encoding the Homo sapiens (Human) cDNA FLJ56323, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_002403.2 |
---|---|
Gene ID (Entrez) | 4255 |
Name | cDNA FLJ56323 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026191
|
enQuireBio™
QP8731-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046191
|
enQuireBio™
QP8731-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036191
|
enQuireBio™
QP8731-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056191
|
enQuireBio™
QP8731-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066191
|
enQuireBio™
QP8731-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076191
|
enQuireBio™
QP8731-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002403.2 | |
cDNA FLJ56323 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4255 | |
Research Use Only | |
MGMT | |
Recombinant Protein | |
E. coli | |
QPAPLERFAS RRPQVLAVRT VCDLVLGKMD KDCEMKRTTL DSPLGKLELS GCEQGLHEIK LLGKGTSAAD AVEVPAPAAV LGGPEPLMQC TAWLNAYFHQ PEAIEEFPVP ALHHPVFQQE SFTRQVLWKL LKVVKFGEVI SYQQLAALAG NPKAARAVGG AMRGNPVPIL IPCHRVVCSS GAVGNYSGGL AVKEWLLAHE GHRLGKPGLG GSSGLAGAWL KGAGATSGSP PAGRN | |
Greater than 85.0% as determined by SDS-PAGE. |