missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CEACAM-5/CD66e Fc Chimera Protein

Product Code. 18796596
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796596 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796596 Supplier R&D Systems™ Supplier No. 10449CM050
  • ONLINE EXCLUSIVE PRICE
    588.29 €
    525.00 € / 50µg
    Save 63.29 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HEK293 Expressed Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human CEACAM-5/CD66e Fc Chimera (Catalog # 10449-CM) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Human Galectin-4 (Catalog # 1227-GA ) binds with an ED50 of 10-90 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_004354.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1048
Molecular Weight (g/mol) M.W.-observed: 135-165 kDa, under reducing conditions, M.W.-theoretical: 96 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human CEACAM-5/CD66e protein Human CEACAM-5/CD66e (Lys35-Ala685) (N-terminus) Accession # NP_004354.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e, CD66e antigen, CEA, CEACAM5, CEACD66e, DKFZp781M2392, Meconium antigen 100
Conjugate Unconjugated
Reconstitution Reconstitute at 400 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein CEACAM5/CD66e
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.