missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEACAM4 Protein

A DNA sequence encoding the Homo sapiens (Human) CEACAM4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5812-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927939

  • 2645.00 € / 1mg
Estimated Shipment: 18-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001808.2
CEACAM4 Protein
Research Use Only
CEACAM4
Recombinant Protein
Yeast
FTIEALPSSA AEGKDVLLLA CNISETIQAY YWHKGKTAEG SPLIAGYITD IQANIPGAAY SGRETVYPNG SLLFQNITLE DAGSYTLRTI NASYDSDQAT GQLHVHQNNV PGLPVGAVAG
Greater than 90% as determined by SDS-PAGE.
1089
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers