missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEACAM6 Protein

A DNA sequence encoding the Homo sapiens (Human) CEACAM6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5813-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987939

  • 2285.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002474.4
CEACAM6 Protein
Research Use Only
CEACAM6
Recombinant Protein
E. coli
KLTIESTPFN VAEGKEVLLL AHNLPQNRIG YSWYKGERVD GNSLIVGYVI GTQQATPGPA YSGRETIYPN ASLLIQNVTQ NDTGFYTLQV IKSDLVNEEA TGQFHVYPEL PKPSISSNNS NPVEDKDAVA FTCEPEVQNT TYLWWVNGQS LPVSPRLQLS NGNMTLTLLS VKRNDAGSYE CEIQNPASAN RSDPVTLNVL YGPDGPTISP SKANYRPGEN LNLSCHAASN PPAQYSWFIN GTFQQSTQEL FIPNITVNNS GSYMCQAHNS ATGLNRTTVT MITVSG
Greater than 90% as determined by SDS-PAGE.
4680
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers