missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CEACAM7 Protein

A DNA sequence encoding the Homo sapiens (Human) CEACAM7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5814-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927949

  • 458.00 € / 100µg
Estimated Shipment: 12-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001278414.1
CEACAM7 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
1087
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CEACAM7
Recombinant Protein
E. coli
TNIDVVPFNV AEGKEVLLVV HNESQNLYGY NWYKGERVHA NYRIIGYVKN ISQENAPGPA HNGRETIYPN GTLLIQNVTH NDAGFYTLHV IKENLVNEEV TRQFYVFSEP PKPSITSNNF NPVENKDIVV LTCQPETQNT TYLWWVNNQS LLVSPRLLLS TDNRTLVLLS ATKNDIGPYE CEIQNPVGAS RSDPVTLNVR YESVQAS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers