missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEBPG / CEBP gamma Protein
A DNA sequence encoding the Homo sapiens (Human) CEBPG / CEBP gamma, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5815-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001239225.1 | |
CEBPG / CEBP gamma Protein | |
Research Use Only | |
CEBPG | |
Recombinant Protein | |
E. coli | |
MSKISQQNST PGVNGISVIH TQAHASGLQQ VPQLVPAGPG GGGKAVAPSK QSKKSSPMDR NSDEYRQRRE RNNMAVKKSR LKSKQKAQDT LQRVNQLKEE NERLEAKIKL LTKELSVLKD LFLEHAHNLA DNVQSISTEN TTADGDNAGQ | |
Greater than 80% as determined by SDS-PAGE. |
1054 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |