missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CECR1 Protein

A cDNA sequence encoding the CECR1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11384-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957098

  • 131.00 € / 2µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CECR1 Protein
Research Use Only
Recombinant Protein
Baculovirus
IDETRAHLLL KEKMMRLGGR LVLNTKEELA NERLMTLKIA EMKEAMRTLI FPPSMHFFQA KHLIERSQVF NILRMMPKGA ALHLHDIGIV TMDWLVRNVT YRPHCHICFT PRGIMQFRFA HPTPRPSEKC SKWILLEDYR KRVQNVTEFD DSLLRNFTLV TQHPEVIYTN QNVVWSKFET IFFTISGLIH YAPVFRDYVF RSMQEFYEDN VLYMEIRARL LPVYELSGEH HDEEWSVKTY QEVAQKFVET HPEFIGIKII YSDHRSKDVA VIAESIRMAM GLRIKFPTVV AGFDLVGHED TGHSLHDYKE ALMIPAKDGV KLPYFFHAGE TDWQGTSIDR NILDALMLNT TRIGHGFALS KHPAVRTYSW KKDIPIEVCP ISNQVLKLVS DLRNHPVATL MATGHPMVIS SDDPAMFGAK GLSYDFYEVF MGIGGMKADL RTLKQLAMNS IKYSTLLESE KNTFMEIWKK RWDKFIADVA TKLEHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CECR1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers