missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CENPA Protein
A cDNA sequence encoding the CENPA was constructed and used to recombinantly synthesize the protein.
131.00 € - 5655.00 €
Specifications
Name | CENPA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947108
|
enQuireBio™
QP11387-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937108
|
enQuireBio™
QP11387-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927108
|
enQuireBio™
QP11387-1MG |
1 mg |
5655.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CENPA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CENPA is supplied in 20mM HEPES buffer pH 8.0, 100mM sodium chloride, and 6M Urea. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MGPRRRSRKP EAPRRRSPSP TPTPGPSRRG PSLGASSHQH SRRRQGWLKE IRKLQKSTHL LIRKLPFSRL AREICVKFTR GVDFNWQAQA LLALQEAAEA FLVHLFEDAY LLTLHAGRVT LFPKDVQLAR RIRGLEEGLG | |
Greater than 90% as determined by SDS-PAGE. |