missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Centrin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7526-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004057.1 | |
Centrin-1 Protein | |
Research Use Only | |
CETN1 | |
Recombinant Protein | |
E. coli | |
MASGFKKPSA ASTGQKRKVA PKPELTEDQK QEVREAFDLF DVDGSGTIDA KELKVAMRAL GFEPRKEEMK KMISEVDREG TGKISFNDFL AVMTQKMSEK DTKEEILKAF RLFDDDETGK ISFKNLKRVA NELGENLTDE ELQEMIDEAD RDGDGEVNEE EFLRIMKKTS LY | |
Greater than 80% as determined by SDS-PAGE. |
1068 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |