missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Centrin-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5821-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001284697.1 | |
Centrin-3 Protein | |
Research Use Only | |
CETN3 | |
Recombinant Protein | |
E. coli | |
MSLALRSELV VDKTKRKKRR ELSEEQKQEI KDAFELFDTD KDEAIDYHEL KVAMRALGFD VKKADVLKIL KDYDREATGK ITFEDFNEVV TDWILERDPH EEILKAFKLF DDDDSGKISL RNLRRVAREL GENMSDEELR AMIEEFDKDG DGEINQEEFI AIMTGDI | |
Greater than 80% as determined by SDS-PAGE. |
1070 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |