missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Citrate synthase / CS Protein
A DNA sequence encoding the Homo sapiens (Human) Citrate synthase / CS, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5873-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004068.2 | |
Citrate synthase / CS Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1431 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CS | |
Recombinant Protein | |
E. coli | |
ASSTNLKDIL ADLIPKEQAR IKTFRQQHGK TVVGQITVDM MYGGMRGMKG LVYETSVLDP DEGIRFRGFS IPECQKLLPK AKGGEEPLPE GLFWLLVTGH IPTEEQVSWL SKEWAKRAAL PSHVVTMLDN FPTNLHPMSQ LSAAVTALNS ESNFARAYAQ GISRTKYWEL IYEDSMDLIA KLPCVAAKIY RNLYREGSGI GAIDSNLDWS HNFTNMLGYT DHQFTELTRL YLTIHSDHEG GNVSAHTSHL VGSALSDPYL SFAAAMNGLA GPLHGLANQE VLVWLTQLQK EVGKDVSDEK LRDYIWNTLN SGRVVPGYGH AVLRKTDPRY TCQREFALKH LPNDPMFKLV AQLYKIVPNV LLEQGKAKNP WPNVDAHSGV LLQYYGMTEM NYYTVLFGVS RALGVLAQLI WSRALGFPLE RPKSMSTEGL MKFVDSK |