missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMBITI Protein
Specifications
Specifications
| Name | CKMBITI Protein |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | 486 IU/mg at 37C. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | P. pastoris |
| Protein Tag | Untagged |
| Buffer | The 7.9 mg/ml protein solution contains 0.02M Potassium Phosphate, 1mM DTT and 50% glycerol, pH 5.0-6.0. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?