missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKMBITI Protein

Product Code. 15934228
Change view
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15934228 10 μg 10µg
15954228 50 μg 50µg
15944228 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15934228 Supplier enQuireBio™ Supplier No. QP1055310ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CKMBITI was constructed and used to recombinantly synthesize the protein.

Specifications

Name CKMBITI Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity 486 IU/mg at 37C.
Product Type Recombinant Protein
Cross Reactivity Human
Species P. pastoris
Protein Tag Untagged
Buffer The 7.9 mg/ml protein solution contains 0.02M Potassium Phosphate, 1mM DTT and 50% glycerol, pH 5.0-6.0.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.