missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMBITII Protein
Specifications
Specifications
| Name | CKMBITII Protein |
| Quantity | 10 μg |
| Regulatory Status | RUO |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | The enzyme activity measured by kinetic assay at 340nm was 883 IU/mg at 37 degrees Celsius, corresponding to a Specific Activity of 1,133ng/ml. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | P. pastoris |
| Protein Tag | Untagged |
| Buffer | Each mg of protein contains 20mM Tris-HCl, pH 6.8, 1mM EDTA and 1mM DTT, 50% (v/v) glycerol. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?