missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CKS2 Protein

Product Code. 15957268
Change view
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15957268 5 μg 5µg
15947268 25 μg 25µg
15937268 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15957268 Supplier enQuireBio™ Supplier No. QP114395ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CKS2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name CKS2 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MASMTGGQQM GRGSHMAHKQ IYYSDKYFDE HYEYRHVMLP RELSKQVPKT HLMSEEEWRRLGVQQSLGWV HYMIHEPEPH ILLFRRPLPK DQQ
Buffer The CKS2 protein solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.