missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKS2 Protein
Specifications
Specifications
| Name | CKS2 Protein |
| Quantity | 25 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | MASMTGGQQM GRGSHMAHKQ IYYSDKYFDE HYEYRHVMLP RELSKQVPKT HLMSEEEWRRLGVQQSLGWV HYMIHEPEPH ILLFRRPLPK DQQ |
| Buffer | The CKS2 protein solution contains 20mM Tris-HCl pH 7.5 and 20% glycerol. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?