missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Clavesin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Clavesin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7684-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040171

  • 505.00 € / 100µg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001010852.2
Clavesin-2 Protein
Research Use Only
CLVS2
Recombinant Protein
E. coli
MTHLQAGLSP ETLEKARLEL NENPDTLHQD IQEVRDMVIT RPDIGFLRTD DAFILRFLRA RKFHHFEAFR LLAQYFEYRQ QNLDMFKSFK ATDPGIKQAL KDGFPGGLAN LDHYGRKILV LFAANWDQSR YTLVDILRAI LLSLEAMIED PELQVNGFVL IIDWSNFTFK QASKLTPSML RLAIEGLQDS FPARFGGIHF VNQPWYIHAL YTVIRPFLKE KTRKRIFLHG NNLNSLHQLI HPEILPSEFG GMLPPYDMGT WARTLLDHEY DDDSEYNVDS YSMPVKEVEK ELSPKSMKRS QSVVDPTVLK RMDKNEEENM QPLLSLD
Greater than 90% as determined by SDS-PAGE.
134829
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers