missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLCF1 Protein
A cDNA sequence encoding the CLCF1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11440-1mg
Additional Details : Weight : 0.01000kg
Specifications
23529 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF | |
Greater than 90% as determined by SDS-PAGE. |
CLCF1 Protein | |
Research Use Only | |
CLCF1 | |
Human | |
His | |
CLCF1 protein solution (1 mg/ml) containing 20mM sodium citrate (pH 3.5), 0.4M Urea and 10% glycerol. |