missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLCF1 Protein

A cDNA sequence encoding the CLCF1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11440-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987268

  • 131.00 € / 5µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

23529
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF
Greater than 90% as determined by SDS-PAGE.
CLCF1 Protein
Research Use Only
CLCF1
Human
His
CLCF1 protein solution (1 mg/ml) containing 20mM sodium citrate (pH 3.5), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers