missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNPY4 Protein

A cDNA sequence encoding the CNPY4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11471-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927368

  • 203.00 € / 10µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

245812
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEFGMLKE EDDDTERLPS KCEVCKLLST ELQAELSRTG RSREVLELGQ VLDTGKRKRH VPYSVSETRL EEALENLCER ILDYSVHAER KGSLRYAKGQ SQTMATLKGL VQKGVKVDLG IPLELWDEPS VEVTYLKKQC ETMLEEFEDI VGDWYFHHQE QPLQNFLCEG HVLPAAETAC LQETWTGKEI TDGEEKTEGE EEQEEEEEEE EEEGGDKMTK TGSHPKLDRE DL
Greater than 85.0% as determined by SDS-PAGE.
CNPY4 Protein
Research Use Only
CNPY4
Human
His
CNPY4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 5mM DTT and 2mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers