missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNPY4 Protein
A cDNA sequence encoding the CNPY4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11471-2ug
Additional Details : Weight : 0.01000kg
Specifications
245812 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFGMLKE EDDDTERLPS KCEVCKLLST ELQAELSRTG RSREVLELGQ VLDTGKRKRH VPYSVSETRL EEALENLCER ILDYSVHAER KGSLRYAKGQ SQTMATLKGL VQKGVKVDLG IPLELWDEPS VEVTYLKKQC ETMLEEFEDI VGDWYFHHQE QPLQNFLCEG HVLPAAETAC LQETWTGKEI TDGEEKTEGE EEQEEEEEEE EEEGGDKMTK TGSHPKLDRE DL | |
Greater than 85.0% as determined by SDS-PAGE. |
CNPY4 Protein | |
Research Use Only | |
CNPY4 | |
Human | |
His | |
CNPY4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 5mM DTT and 2mM EDTA. |