missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COA1 Protein
A DNA sequence encoding the Homo sapiens (Human) COA1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5738-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001308126.1 | |
COA1 Protein | |
Research Use Only | |
COA1 | |
Recombinant Protein | |
E. coli | |
QKFHSRALYY KLAVEQLQSH PEAQEALGPP LNIHYLKLID RENFVDIVDA KLKIPVSGSK SEGLLYVHSS RGGPFQRWHL DEVFLELKDG QQIPVFKLSG ENGDEVKKE | |
Greater than 90% as determined by SDS-PAGE. |
55744 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |