missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen-III Protein
A cDNA sequence encoding the Collagen-III was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11479-100mg
Additional Details : Weight : 0.01000kg
Specifications
1281 | |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Pichia Pastoris | |
MMSFVQKGSW LLLALLHPTI ILAQQEAVEG GCSHLGQSYA DRDVWKPEPC QICVCDSGSV LCDDIICDDQ ELDCPNPEIP FGECCAVCPQ PPTAPTRPPN GQGPQGPKGD PGPPGIPGRN GDPGIPGQPG SPGSPGPPGI CESCPTGPQN YSPQYDSYDV KSGVAVGGLA GYPGPAGPPG PPGPPGTSGH PGSPGSPGYQ GPPGEPGQAG PSGPPGPPGA IGPSGPAGKD GESGRPGRPG ERGLPGPPGI KGPAGIPGFP GMKGHRGFDG RNGEKGETGA PGLKGENGLP GENGAPGPMG PRGAPGERGR PGLPGAAGAR GNDGARGSDG QPGPPGPPGT AGFPGSPGAK GEVGPAGSPG SNGAPGQRGE PGPQGHAGAQ GPPGPPGING SPGGKGEMGP AGIPGAPGLM GARGPPGPAG ANGAPGLRGG AGEPGKNGAK GEPGPRGERG EAGIPGVPGA KGEDGKDGSP GEPGANGLPG AAGERGAPGF RGPAGPNGIP GEKGPAGERG APGPAGPRGA AGEPGRDGVP GGPGMRGMPG SPGGPGSDGK PGPPGSQGES GRPGPPGPSG PRGQPGVMGF PGPKGNDGAP GKNGERGGPG GPGPQGPPGK NGETGPQGPP GPTGPGGDKG DTGPPGPQGL QGLPGTGGPP GENGKPGEPG PKGDAGAPGA PGGKGDAGAP GERGPPGLAG APGLRGGAGP PGPEGGKGAA GPPGPPGAAG TPGLQGMPGE RGGLGSPGPK GDKGEPGGPG ADGVPGKDGP RGPTGPIGPP GPAGQPGDKG EGGAPGLPGI AGPRGSPGER GETGPPGPAG FPGAPGQNGE PGGKGERGAP GEKGEGGPPG VAGPPGGSGP AGPPGPQGVK GERGSPGGPG AAGFPGARGL PGPPGSNGNP GPPGPSGSPG KDGPPGPAGN TGAPGSPGVS GPKGDAGQPG EKGSPGAQGP PGAPGPLGIA GITGARGLAG PPGMPGPRGS PGPQGVKGES GKPGANGLSG ERGPPGPQGL PGLAGTAGEP GRDGNPGSDG LPGRDGSPGG KGDRGENGSP GAPGAPGHPG PPGPVGPAGK SGDRGESGPA GPAGAPGPAG SRGAPGPQGP RGDKGETGER GAAGIKGHRG FPGNPGAPGS PGPAGQQGAI GSPGPAGPRG PVGPSGPPGK DGTSGHPGPI GPPGPRGNRG ERGSEGSPGH PGQPGPPGPP GAPGPCCGGV GAAAIAGIGG EKAGGFAPYY GDEPMDFKIN TDEIMTSLKS VNGQIESLIS PDGSRKNPAR NCRDLKFCHP ELKSGEYWVD PNQGCKLDAI KVFCNMETGE TCISANPLNV PRKHWWTDSS AEKKHVWFGE SMDGGFQFSY GNPELPEDVL DVHLAFLRLL SSRASQNITY HCKNSIAYMD QASGNVKKAL KLMGSNEGEF KAEGNSKFTY TVLEDGCTKH TGEWSKTVFE YRTRKAVRLP IVDIAPYDIG GPDQEFGVDV GPVCFL |
Collagen-III Protein | |
Research Use Only | |
COL3A1 | |
Human | |
Untagged | |
The Human Collagen III solution in 10mM HCl. |