missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMMD7 Protein
A cDNA sequence encoding the COMMD7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11481-20ug
Additional Details : Weight : 0.01000kg
Specifications
COMMD7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGRLHCT EDPVPEAVGG DMQQLNQLGA QQFSALTEVL FHFLTEPKEV ERFLAQLSEF ATTNQISLGS LRSIVKSLLL VPNGALKKSL TAKQVQADFI TLGLSEEKAT YFSEKWKQNA PTLARWAIGQ TLMINQLIDM EWKFGVTSGS SELEKVGSIF LQLKLVVKKG NQTENVYIEL TLPQFYSFLH EMERVRTSME CFC | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
COMMD7 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |