missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human COQ9 Protein

A cDNA sequence encoding the COQ9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11488-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927418

  • 3595.00 € / 1mg
Estimated Shipment: 12-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

COQ9 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
COQ9 protein solution (1 mg/ml) in Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSRSSDEQK QQPPNSFSQQ HSETQGAEKP DPESSHSPPR YTDQGGEEEE DYESEEQLQH RILTAALEFV PAHGWTAEAI AEGAQSLGLS SAAASMFGKD GSELILHFVT QCNTRLTRVL EEEQKLVQLG QAEKRKTDQF LRDAVETRLR MLIPYIEHWP RALSILMLPH NIPSSLSLLT SMVDDMWHYA GDQSTDFNWY TRRAMLAAIY NTTELVMMQD SSPDFEDTWR FLENRVNDAM NMGHTAKQVK STGEALVQGL MGAAVTLKNL TGLNQRR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers