missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRABP2 Protein
A cDNA sequence encoding the CRABP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11498-1mg
Additional Details : Weight : 0.01000kg
Specifications
CRABP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPNFSGNWKI IRSENFEELL KVLGVNVMLR KIAVAAASKP AVEIKQEGDT FYIKTSTTVR TTEINFKVGE EFEEQTVDGR PCKSLVKWES ENKMVCEQKL LKGEGPKTSW TRELTNDGEL ILTMTADDVV CTRVYVRE | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The CRABP2 protein solution contains 20mM Tris-HCl pH 8 and 20% glycerol. |