missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRABP2 Protein

A cDNA sequence encoding the CRABP2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11498-5ug

 View more versions of this product

Product Code. 15947448

  • 131.00 € / 5µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CRABP2 Protein
Research Use Only
Recombinant Protein
E. coli
MPNFSGNWKI IRSENFEELL KVLGVNVMLR KIAVAAASKP AVEIKQEGDT FYIKTSTTVR TTEINFKVGE EFEEQTVDGR PCKSLVKWES ENKMVCEQKL LKGEGPKTSW TRELTNDGEL ILTMTADDVV CTRVYVRE
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The CRABP2 protein solution contains 20mM Tris-HCl pH 8 and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers