missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRCP Protein
A cDNA sequence encoding the CRCP was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | CRCP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15977448
|
enQuireBio™
QP11499-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967448
|
enQuireBio™
QP11499-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957448
|
enQuireBio™
QP11499-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CRCP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CRCP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEVKDANSAL LSNYEVFQLL TDLKEQRKES GKNKHSSGQQ NLNTITYETL KYISKTPCRH QSPEIVREFL TALKSHKLTK AEKLQLLNHR PVTAVEIQLM VEESEERLTE EQIEALLHTV TSILPAEPEA EQKKNTNSNV AMDEEDPA | |
Greater than 95% as determined by SDS-PAGE. |