missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Creatine Kinase BB Protein

Product Code. 18788577
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18788577 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788577 Supplier R&D Systems™ Supplier No. 9076CK050
  • ONLINE EXCLUSIVE PRICE
    419.48 €
    373.80 € / 50µg
    Save 45.68 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to phosphorylate creatine. The specific activity is >3,500 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12277
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and TCEP.
Gene ID (Entrez) 1152
Molecular Weight (g/mol) M.W. (Observed): 43 kDa, reducing conditions; M.W. (theoretical): 43 kDa
Quantity 50 μg
Source E. coli-derived human Creatine Kinase BB protein Pro2 - Lys381, with a N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Common Name Creatine Kinase BB
Gene Symbol CKB
Biological Activity Measured by its ability to phosphorylate creatine. The specific activity is >3,500 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System E. coli-derived Pro2 - Lys381, with a N-terminal Met and 6-His tag
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Creatine Kinase BB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.