missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRIPT Protein
A cDNA sequence encoding the CRIPT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11504-2ug
Additional Details : Weight : 0.01000kg
Specifications
CRIPT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSIHHHHHH SSGLVPRGSH MGSMVCEKCE KKLGTVITPD TWKDGARNTT ESGGRKLNEN KALTSKKARF DPYGKNKFST CRICKSSVHQ PGSHYCQGCA YKKGICAMCG KKVLDTKNYK QTSV | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CRIPT solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |