missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRNN Protein
A cDNA sequence encoding the CRNN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11510-25ug
Additional Details : Weight : 0.01000kg
Specifications
CRNN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPQLLQNING IIEAFRRYAR TEGNCTALTR GELKRLLEQE FADVIVKPHD PATVDEVLRL LDEDHTGTVE FKEFLVLVFK VAQACFKTLS ESAEGACGSQ ESGSLHSGAS QELGEGQRSG TEVGRAGKGQ HYEGSSHRQS QQGSRGQNRP GVQTQGQATG SAWVSSYDRQ AESQSQERIS PQIQLSGQTE QTQKAGEGKR NQTTEMRPER QPQTREQDRA HQTGETVTGS GTQTQAGATQ TVEQDSSHQT GRTSKQTQEA TNDQNRGTET HGQGRSQTSQ AVTGGHAQIQ AGTHTQTPTQ TVEQDSSHQT GSTSTQTQES TNGQNRGTEI HGQGRSQTSQ AVTGGHTQIQ AGSHTETVEQ DRSQTVSHGG AREQGQTQTQ PGSGQRWMQV SNPEAGETVP GGQAQTGAST EPGRQEWSST HPRRCVTEGQ GDRQPTVVGE EWVDDHSRET VILRLDQGNL HTSVSSAQGQ DAAQSEEKRG ITARELYSYL RSTKP | |
Greater than 85% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CRNN solution contains 20mM Tris-HCl pH 8 and 10% glycerol. |