missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRTAM Protein

A cDNA sequence encoding the CRTAM was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11515-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957498

  • 7540.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

56253
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
SLTNHTETIT VEEGQTLTLK CVTSLRKNSS LQWLTPSGFT IFLNEYPALK NSKYQLLHHS ANQLSITVPN VTLQDEGVYK CLHYSDSVST KEVKVIVLAT PFKPILEASV IRKQNGEEHV VLMCSTMRSK PPPQITWLLG NSMEVSGGTL HEFETDGKKC NTTSTLIIHT YGKNSTVDCI IRHRGLQGRK LVAPFRFEDL VTDEETASDA LERNSLSSQD PQQPTSTVSV TEDSSTSEID KEEKEQTTQD PDLTTEANPQ YLGLARKKSG LEHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
CRTAM Protein
Research Use Only
CRTAM
Human
His
CRTAM protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers