missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRTAP Protein
A cDNA sequence encoding the CRTAP was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 10491 |
---|---|
Name | CRTAP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CRTAP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15997498
|
enQuireBio™
QP11516-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987498
|
enQuireBio™
QP11516-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977498
|
enQuireBio™
QP11516-100UG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10491 | |
Research Use Only | |
CRTAP | |
Human | |
His | |
The CRTAP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
CRTAP Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQYERYSF RSFPRDELMP LESAYRHALD KYSGEHWAES VGYLEISLRL HRLLRDSEAF CHRNCSAAPQ PEPAAGLASY PELRLFGGLL RRAHCLKRCK QGLPAFRQSQ PSREVLADFQ RREPYKFLQF AYFKANNLPK AIAAAHTFLL KHPDDEMMKR NMAYYKSLPG AEDYIKDLET KSYESLFIRA VRAYNGENWR TSITDMELAL PDFFKAFYEC LAACEGSREI KDFKDFYLSI ADHYVEVLEC KIQCEENLTP VIGGYPVEKF VATMYHYLQF AYYKLNDLKN AAPCAVSYLL FDQNDKVMQQ NLVYYQYHRD TWGLSDEHFQ PRPEAVQFFN VTTLQKELYD FAKENIMDDD EGEVVEYVDD LLELEETS | |
Greater than 85% as determined by SDS-PAGE. |